Assay Details
Target Gene Details
Entrez Gene ID: | 14702 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 2 |
Gene Aliases: |
82 |
Location: |
Chr.14:19872558-19977462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng2 | NM_001038637.1 | NP_001033726.1 | ||
| NM_001285908.1 | NP_001272837.1 | |||
| NM_001285909.1 | NP_001272838.1 | |||
| NM_001285910.1 | NP_001272839.1 | |||
| NM_001285911.1 | NP_001272840.1 | |||
| NM_010315.4 | NP_034445.1 | |||
| AK003588.1 | BAB22878.1 | |||
| AK012405.1 | BAB28219.1 | |||
| AK036138.1 | BAC29316.1 | |||
| AK075698.1 | BAC35896.1 | |||
| AK158697.1 | BAE34615.1 | |||
| AK160396.1 | BAE35765.1 | |||
| BC021599.1 | AAH21599.1 | |||
| BY108799.1 | ||||
| BY245532.1 | ||||
| CA324196.1 | ||||
| W48309.1 |
More Information
Set Membership: |
|

Genomic Map