Assay Details
Target Gene Details
Entrez Gene ID: | 14714 |
Gene Name: | gonadotropin releasing hormone 1 |
Gene Aliases: |
Gnrh, Gnrh2, LHRH, Lhrh1, Lnrh, hpg |
Location: |
Chr.14:67745229-67749446 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnrh1 | NM_008145.2 | NP_032171.1 | ||
| CX763652.1 |
More Information
Set Membership: |
|

Genomic Map