Assay Details
Target Gene Details
Entrez Gene ID: | 75088 |
Gene Name: | RIKEN cDNA 4930505G20 gene |
Gene Aliases: |
- |
Location: |
Chr.14:115396551-115409552 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930505G20Rik | NR_045761.1 | |||
| AK015702.1 |
Target Gene Details
Entrez Gene ID: | 103978 |
Gene Name: | glypican 5 |
Gene Aliases: |
A230034F01Rik |
Location: |
Chr.14:115092212-116525192 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpc5 | NM_175500.4 | NP_780709.1 | ||
| AF001463.1 | AAC12262.1 | |||
| AK038547.1 | BAC30037.1 | |||
| AK084223.1 | BAC39143.1 | |||
| BC115712.1 |
More Information
Set Membership: |
|

Genomic Map