Assay Details
Target Gene Details
Entrez Gene ID: | 628705 |
Gene Name: | PHD finger protein 11C |
Gene Aliases: |
EG628705, Gm6907, Phf11 |
Location: |
Chr.14:59380833-59440890 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phf11c | NM_001164289.1 | NP_001157761.1 | ||
| EU155106.1 |
More Information
Set Membership: |
|

Genomic Map