Assay Details
Target Gene Details
Entrez Gene ID: | 110168 |
Gene Name: | G protein-coupled receptor 18 |
Gene Aliases: |
- |
Location: |
Chr.14:121911434-121915774 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr18 | NM_182806.1 | NP_877958.1 | ||
| AK156039.1 | 1 | 129 | BAE33557.1 | |
| AK156868.1 | 1 | 121 | BAE33880.1 | |
| AK157029.1 | 1 | 64 | BAE33938.1 | |
| AK172401.1 | BAE42987.1 | |||
| BC034872.1 | AAH34872.1 |
Target Gene Details
Entrez Gene ID: | 68889 |
Gene Name: | ubiquitin associated domain containing 2 |
Gene Aliases: |
1190008A14Rik, AW554959, Phgdhl1 |
Location: |
Chr.14:121878556-122021035 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubac2 | NM_026861.2 | NP_081137.2 | ||
| AK035384.1 | ||||
| AK049109.1 | BAC33546.1 | |||
| AK158177.1 | BAE34394.1 | |||
| AK199665.1 | ||||
| BC024467.1 | AAH24467.1 |
More Information
Set Membership: |
|

Genomic Map