Assay Details
Target Gene Details
Entrez Gene ID: | 13850 |
Gene Name: | epoxide hydrolase 2, cytoplasmic |
Gene Aliases: |
CEH, Eph2, SEH, sEP |
Location: |
Chr.14:66084372-66124522 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephx2 | NM_001271402.1 | 1 | 108 | NP_001258331.1 |
| NM_007940.4 | 1 | 108 | NP_031966.2 | |
| AK002415.1 | 1 | 78 | ||
| AK142714.1 | 1 | 81 | BAE25172.1 | |
| BC015087.1 | 1 | 84 | AAH15087.1 | |
| L05781.1 | AAA37555.1 | |||
| Z37107.1 | CAA85471.1 |
More Information
Set Membership: |
|

Genomic Map