Assay Details
Target Gene Details
Entrez Gene ID: | 13850 |
Gene Name: | epoxide hydrolase 2, cytoplasmic |
Gene Aliases: |
CEH, Eph2, SEH, sEP |
Location: |
Chr.14:66084372-66124522 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephx2 | NM_001271402.1 | NP_001258331.1 | ||
| NM_001271403.1 | NP_001258332.1 | |||
| NM_001271421.1 | NP_001258350.1 | |||
| NM_007940.4 | NP_031966.2 | |||
| AK002415.1 | ||||
| AK142714.1 | BAE25172.1 | |||
| AY098585.1 | AAM28238.1 | |||
| BC015087.1 | AAH15087.1 | |||
| EF597241.1 | ||||
| L05781.1 | AAA37555.1 | |||
| Z37107.1 | CAA85471.1 |
More Information
Set Membership: |
|

Genomic Map