Assay Details
Target Gene Details
Entrez Gene ID: | 73977 |
Gene Name: | RIKEN cDNA 4930438E09 gene |
Gene Aliases: |
- |
Location: |
Chr.14:67332042-67397742 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930438E09Rik | NR_045961.1 | |||
| AK015336.1 |
Target Gene Details
Entrez Gene ID: | 13592 |
Gene Name: | early B cell factor 2 |
Gene Aliases: |
D14Ggc1e, EBF-2, Mmot1, O/E-3, OE-3 |
Location: |
Chr.14:67233292-67430952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ebf2 | NM_001276387.1 | NP_001263316.1 | ||
| NM_010095.6 | NP_034225.1 | |||
| AK053013.1 | BAC35239.1 | |||
| AK139262.1 | BAE23936.1 | |||
| BC049188.1 | AAH49188.1 | |||
| BC050922.1 | AAH50922.1 | |||
| U71189.1 | AAB58423.1 | |||
| U82441.1 | AAC64322.1 | |||
| U92703.1 | AAB58323.1 |
More Information
Set Membership: |
|

Genomic Map