Assay Details
Target Gene Details
Entrez Gene ID: | 65246 |
Gene Name: | exportin 7 |
Gene Aliases: |
4930506C02Rik, BB164534, Ranbp16, mKIAA0745 |
Location: |
Chr.14:70654246-70766634 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xpo7 | NM_023045.2 | 7 | 776 | NP_075532.1 |
| AJ297360.1 | 7 | 682 | CAC17621.1 | |
| AK028303.1 | 5 | 521 | BAC25870.1 | |
| AK035775.1 | 7 | 701 | BAC29182.1 | |
| AK036837.1 | 5 | 521 | BAC29600.1 | |
| AK051105.1 | 7 | 698 | ||
| AK122361.1 | 6 | 661 | BAC65643.1 | |
| AK143729.1 | 7 | 745 | ||
| AK164595.1 | 7 | 704 | BAE37843.1 | |
| BC029702.1 | 7 | 702 | AAH29702.1 |
More Information
Set Membership: |
|

Genomic Map