Assay Details
Target Gene Details
Entrez Gene ID: | 66242 |
Gene Name: | mitochondrial ribosomal protein S16 |
Gene Aliases: |
1500011E11Rik, Mprs16 |
Location: |
Chr.14:20391231-20393555 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps16 | NM_025440.2 | NP_079716.1 | ||
| AB049949.1 | BAB41002.1 | |||
| AK005206.1 | BAB23884.1 |
More Information
Set Membership: |
|

Genomic Map