Assay Details
Target Gene Details
Entrez Gene ID: | 219024 |
Gene Name: | transmembrane protein 55b |
Gene Aliases: |
BC039161 |
Location: |
Chr.14:50926070-50930905 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem55b | NM_001033271.4 | 3 | 489 | NP_001028443.1 |
| AK144251.1 | 3 | 489 | BAE25798.1 | |
| AK152640.1 | 3 | 475 | ||
| AK152658.1 | 3 | 475 | ||
| AK159641.1 | 3 | 493 | BAE35254.1 | |
| AK169026.1 | 2 | 914 | ||
| BC106163.1 | 3 | 468 | AAI06164.1 |

Genomic Map