Assay Details
Target Gene Details
Entrez Gene ID: | 68043 |
Gene Name: | N-6 adenine-specific DNA methyltransferase 2 (putative) |
Gene Aliases: |
2510005D08Rik, AW045965, Ayu21-96, Gt(pU21)96Imeg, GtAyu21-96 |
Location: |
Chr.14:57549596-57571569 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N6amt2 | NM_026526.2 | NP_080802.1 | ||
| AK005312.1 | BAB23947.1 | |||
| AK010911.1 | BAB27262.1 | |||
| AK012083.1 | BAB28017.1 | |||
| BC051925.1 | AAH51925.1 |

Genomic Map