Assay Details
Target Gene Details
Entrez Gene ID: | 69721 |
Gene Name: | NFKB inhibitor interacting Ras-like protein 1 |
Gene Aliases: |
2400004O09Rik, KBRAS1, kappaB-Ras1 |
Location: |
Chr.14:18271142-18284003 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkiras1 | NM_023526.3 | NP_076015.2 | ||
| AF225707.1 | AAF43150.1 | |||
| AK010285.1 | ||||
| AK028539.1 | BAC25998.1 | |||
| AK148040.1 | BAE28306.1 | |||
| AK167340.1 | BAE39441.1 | |||
| BC061092.1 | AAH61092.1 | |||
| BY268943.1 |

Genomic Map