Assay Details
Target Gene Details
Entrez Gene ID: | 20618 |
Gene Name: | synuclein, gamma |
Gene Aliases: |
C79089, persyn |
Location: |
Chr.14:34370274-34374669 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sncg | NM_011430.3 | NP_035560.1 | ||
| AA690669.1 | ||||
| AF017255.1 | AAC98893.1 | |||
| AJ786826.1 | CAH17958.1 | |||
| AK141509.1 | BAE24706.1 | |||
| BC028508.1 | AAH28508.1 |
More Information
Set Membership: |
|

Genomic Map