Assay Details
Target Gene Details
Entrez Gene ID: | 70021 |
Gene Name: | 5'-nucleotidase domain containing 2 |
Gene Aliases: |
2510015F01Rik |
Location: |
Chr.14:31131058-31139124 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5dc2 | NM_027289.1 | NP_081565.1 | ||
| BC011230.1 | AAH11230.1 |
More Information
Set Membership: |
|

Genomic Map