Assay Details
Target Gene Details
Entrez Gene ID: | 21974 |
Gene Name: | topoisomerase (DNA) II beta |
Gene Aliases: |
D230016L12Rik, Top-2 |
Location: |
Chr.14:16365206-16430787 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top2b | NM_009409.2 | 21 | 3130 | NP_033435.2 |
| AK194939.1 | ||||
| AK194992.1 | ||||
| BC041106.1 | 21 | 3150 | AAH41106.1 | |
| BC054541.1 | 21 | 3130 | AAH54541.1 | |
| D38046.1 | 21 | 2703 | BAA07236.1 |
More Information
Set Membership: |
|

Genomic Map