Assay Details
Target Gene Details
Entrez Gene ID: | 21974 |
Gene Name: | topoisomerase (DNA) II beta |
Gene Aliases: |
D230016L12Rik, Top-2 |
Location: |
Chr.14:16365206-16430787 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top2b | NM_009409.2 | NP_033435.2 | ||
| AK041054.1 | BAC30801.1 | |||
| AK087535.1 | ||||
| AK163043.1 | BAE37168.1 | |||
| BC041106.1 | AAH41106.1 | |||
| BC054541.1 | AAH54541.1 | |||
| D38046.1 | BAA07236.1 |
More Information
Set Membership: |
|

Genomic Map