Assay Details
Target Gene Details
Entrez Gene ID: | 74551 |
Gene Name: | phosphoenolpyruvate carboxykinase 2 (mitochondrial) |
Gene Aliases: |
1810010O14Rik, 9130022B02Rik, PEPCK-M |
Location: |
Chr.14:55540263-55550777 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pck2 | NM_028994.2 | 1 | 166 | NP_083270.1 |
| AK030501.1 | 1 | 167 | BAC26991.1 | |
| AK031612.1 | BAC27477.1 | |||
| AK034927.1 | 1 | 45 | BAC28883.1 | |
| AK147972.1 | BAE28259.1 | |||
| AK161944.1 | 1 | 46 | ||
| AK165094.1 | 1 | 44 | ||
| AK170242.1 | BAE41656.1 | |||
| BC010318.1 | AAH10318.1 |
More Information
Set Membership: |
|

Genomic Map