Assay Details
Target Gene Details
Entrez Gene ID: | 27374 |
Gene Name: | protein arginine N-methyltransferase 5 |
Gene Aliases: |
Jbp1, Skb1 |
Location: |
Chr.14:54507182-54517470 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prmt5 | NM_013768.3 | NP_038796.2 | ||
| AF167573.1 | AAF04503.1 | |||
| AK076346.1 | ||||
| AK132414.1 | BAE21155.1 | |||
| AK165165.1 | BAE38057.1 | |||
| AK206591.1 | ||||
| AK207153.1 | ||||
| AK207622.1 | ||||
| AK207926.1 | ||||
| AK210265.1 | ||||
| AK210298.1 | ||||
| AK210921.1 | ||||
| AK212522.1 | ||||
| AK214114.1 | ||||
| AK214487.1 | ||||
| BC023905.1 | AAH23905.1 |
More Information
Set Membership: |
|

Genomic Map