Assay Details
Target Gene Details
Entrez Gene ID: | 223776 |
Gene Name: | selenoprotein O |
Gene Aliases: |
1300018J18Rik |
Location: |
Chr.15:89089107-89100340 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selo | NM_027905.2 | 2 | 621 | NP_082181.2 |
| AK005048.1 | 2 | 621 | BAB23774.1 | |
| AK054432.1 | 2 | 641 | ||
| AK088096.1 | 2 | 646 | ||
| AK161059.1 | 2 | 630 | ||
| BC117817.1 | 3 | 136 | ||
| BC141293.1 | 2 | 690 |

Genomic Map