Assay Details
Target Gene Details
Entrez Gene ID: | 223776 |
Gene Name: | selenoprotein O |
Gene Aliases: |
1300018J18Rik |
Location: |
Chr.15:89089107-89100340 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selo | NM_027905.2 | NP_082181.2 | ||
| AK005048.1 | BAB23774.1 | |||
| AK054432.1 | ||||
| AK088096.1 | ||||
| AK161059.1 | ||||
| BC117817.1 | ||||
| BC141293.1 |

Genomic Map