Assay Details
Target Gene Details
Entrez Gene ID: | 223776 |
Gene Name: | selenoprotein O |
Gene Aliases: |
1300018J18Rik |
Location: |
Chr.15:89089107-89100340 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selo | NM_027905.2 | 6 | 1433 | NP_082181.2 |
| AK005048.1 | 6 | 1433 | BAB23774.1 | |
| BC048849.1 | 3 | 456 | AAH48849.1 | |
| BC117817.1 | 7 | 948 | ||
| BC141293.1 | 6 | 1502 |

Genomic Map