Assay Details
Target Gene Details
Entrez Gene ID: | 11482 |
Gene Name: | activin A receptor, type II-like 1 |
Gene Aliases: |
AI115505, AI427544, Acvrlk1, Alk1 |
Location: |
Chr.15:101128522-101145336 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvrl1 | NM_001277255.1 | NP_001264184.1 | ||
| NM_001277257.1 | NP_001264186.1 | |||
| NM_001277258.1 | NP_001264187.1 | |||
| NM_001277259.1 | NP_001264188.1 | |||
| NM_009612.3 | NP_033742.2 | |||
| L48015.1 | AAB03642.1 |
More Information
Set Membership: |
|

Genomic Map