Assay Details
Target Gene Details
Entrez Gene ID: | 11482 |
Gene Name: | activin A receptor, type II-like 1 |
Gene Aliases: |
AI115505, AI427544, Acvrlk1, Alk1 |
Location: |
Chr.15:101128522-101145336 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvrl1 | NM_001277255.1 | NP_001264184.1 | ||
| NM_001277257.1 | NP_001264186.1 | |||
| NM_001277258.1 | NP_001264187.1 | |||
| NM_001277259.1 | NP_001264188.1 | |||
| NM_009612.3 | NP_033742.2 | |||
| AK134712.1 | ||||
| AK160915.1 | BAE36089.1 | |||
| AK170870.1 | BAE42083.1 | |||
| BB869400.1 | ||||
| BB869983.1 | ||||
| BC014291.1 | AAH14291.1 | |||
| BC015083.1 | AAH15083.1 | |||
| CT010330.1 | CAJ18538.1 | |||
| L48015.1 | AAB03642.1 | |||
| Z31664.1 | CAA83484.1 |
More Information
Set Membership: |
|

Genomic Map