Assay Details
Target Gene Details
Entrez Gene ID: | 69089 |
Gene Name: | oxidase assembly 1-like |
Gene Aliases: |
1810020M02Rik, AI894287 |
Location: |
Chr.14:54360841-54369669 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxa1l | NM_026936.3 | NP_081212.1 | ||
| AK007567.1 | BAB25113.1 | |||
| AK028379.1 | ||||
| AK044817.1 | BAC32104.1 | |||
| AK052432.1 | BAC34985.1 | |||
| AK077624.1 | BAC36908.1 | |||
| AK087756.1 | ||||
| AK139484.1 | BAE24033.1 | |||
| AK154304.1 | BAE32503.1 | |||
| AK154532.1 | BAE32657.1 | |||
| AK170898.1 | BAE42101.1 | |||
| AK214867.1 | ||||
| BC027191.1 | AAH27191.1 |
More Information
Set Membership: |
|

Genomic Map