Assay Details
Target Gene Details
Entrez Gene ID: | 66656 |
Gene Name: | eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein) |
Gene Aliases: |
EF-1-delta |
Location: |
Chr.15:75894796-75909556 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef1d | NM_001285429.1 | NP_001272358.1 | ||
| NM_001285430.1 | NP_001272359.1 | |||
| NM_001285431.1 | NP_001272360.1 | |||
| NM_001285432.1 | NP_001272361.1 | |||
| NM_001285433.1 | NP_001272362.1 | |||
| NM_001285434.1 | NP_001272363.1 | |||
| NM_023240.3 | NP_075729.2 | |||
| NM_029663.2 | NP_083939.1 | |||
| AF304351.1 | AAG17466.1 | |||
| AK010347.1 | BAB26870.1 | |||
| AK017616.1 | BAB30841.1 | |||
| AK017796.1 | ||||
| AK044934.1 | BAC32149.1 | |||
| AK054335.1 | BAC35736.1 | |||
| BC013059.1 | AAH13059.1 | |||
| BC079855.1 | AAH79855.1 | |||
| BQ177630.1 | ||||
| BY035199.1 | ||||
| BY104511.1 | ||||
| BY154960.1 | ||||
| DV062370.1 |
More Information
Set Membership: |
|

Genomic Map