Assay Details
Target Gene Details
Entrez Gene ID: | 80910 |
Gene Name: | G protein-coupled receptor 84 |
Gene Aliases: |
EX33 |
Location: |
Chr.15:103308233-103311242 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr84 | NM_030720.1 | 1 | 67 | NP_109645.1 |
| AF272948.1 | 1 | 67 | AAK01859.1 | |
| BC023249.1 | 1 | 76 | AAH23249.1 |
More Information
Set Membership: |
|

Genomic Map