Assay Details
Target Gene Details
Entrez Gene ID: | 93696 |
Gene Name: | chromatin accessibility complex 1 |
Gene Aliases: |
2410152E03Rik, 2810406L04Rik, YCL1 |
Location: |
Chr.15:73090412-73094075 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrac1 | NM_053068.3 | 1 | 84 | NP_444298.1 |
| AF230805.1 | AAF67145.1 | |||
| AK010812.1 | 1 | 85 | BAB27197.1 | |
| BC016593.1 | AAH16593.1 | |||
| BC061043.1 | 1 | 83 | AAH61043.1 |
More Information
Set Membership: |
|

Genomic Map