Assay Details
Target Gene Details
Entrez Gene ID: | 18606 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 2 |
Gene Aliases: |
ATX, Npps2, PD-Ialpha, Pdnp2, lysoPLD |
Location: |
Chr.15:54838679-54952887 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Enpp2 | NM_001136077.2 | NP_001129549.1 | ||
| NM_001285994.1 | NP_001272923.1 | |||
| NM_001285995.1 | NP_001272924.1 | |||
| NM_015744.3 | NP_056559.2 | |||
| AF123542.1 | AAD46480.1 | |||
| AK038940.1 | ||||
| AK088491.1 | ||||
| AK161144.1 | BAE36214.1 | |||
| AK163881.1 | ||||
| BC003264.1 | AAH03264.1 | |||
| BC058759.1 | AAH58759.1 | |||
| EU131009.1 | ||||
| EU131010.1 |

Genomic Map