Assay Details
Target Gene Details
Entrez Gene ID: | 14663 |
Gene Name: | glycosylation dependent cell adhesion molecule 1 |
Gene Aliases: |
MC26, Sgp50, glyCAM-1 |
Location: |
Chr.15:103562759-103565081 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glycam1 | NM_001289587.1 | 3 | 272 | NP_001276516.1 |
| NM_008134.3 | 3 | 272 | NP_032160.1 | |
| AK021358.1 | 3 | 270 | BAB32385.1 | |
| BC145548.1 | 3 | 239 | ||
| BC145832.1 | 3 | 245 | ||
| BC145858.1 | 3 | 245 | ||
| BE947771.1 | ||||
| M93428.1 | 3 | 244 | AAA37710.1 |

Genomic Map