Assay Details
Target Gene Details
Entrez Gene ID: | 12181 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
AU020183, AW146150, D18861, Erb1p, Kiaa0124, mKIAA0124 |
Location: |
Chr.15:76452996-76477269 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bop1 | NM_013481.1 | 4 | 474 | NP_038509.1 |
| AK002984.1 | ||||
| AK079627.1 | 4 | 454 | BAC37708.1 | |
| AK129058.1 | 4 | 447 | BAC97868.1 | |
| AK144095.1 | 4 | 485 | ||
| AK167110.1 | 4 | 457 | BAE39258.1 | |
| AK167225.1 | 4 | 458 | BAE39350.1 | |
| AK167829.1 | 4 | 452 | BAE39851.1 | |
| AK167878.1 | 4 | 462 | ||
| AK168127.1 | 4 | 455 | BAE40096.1 | |
| AK180635.1 | ||||
| AK181923.1 | ||||
| AK187881.1 | ||||
| AK199312.1 | ||||
| AK201242.1 | ||||
| AK206410.1 | ||||
| BC012693.1 | 4 | 448 | AAH12693.1 | |
| U77415.1 | 4 | 474 | AAB19223.1 |
More Information
Set Membership: |
|

Genomic Map