Assay Details
Target Gene Details
Entrez Gene ID: | 73739 |
Gene Name: | chibby homolog 1 (Drosophila) |
Gene Aliases: |
1110014P06Rik, CBY, PGEA14, Pgea1, arb1 |
Location: |
Chr.15:79659166-79667662 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cby1 | NM_028634.3 | 4 | 378 | NP_082910.1 |
| AF331040.1 | 4 | 378 | AAL56061.1 | |
| AK003719.1 | 4 | 354 | BAB22956.1 | |
| BC005733.1 | 3 | 331 | AAH05733.1 |

Genomic Map