Assay Details
Target Gene Details
Entrez Gene ID: | 110542 |
Gene Name: | anti-Mullerian hormone type 2 receptor |
Gene Aliases: |
Misiir, Misrii, Mrii |
Location: |
Chr.15:102445028-102454640 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amhr2 | NM_144547.2 | NP_653130.2 | ||
| AF503863.1 | AAM28608.1 | |||
| BC119571.1 |
More Information
Set Membership: |
|

Genomic Map