Assay Details
Target Gene Details
Entrez Gene ID: | 100048895 |
Gene Name: | RIKEN cDNA 9330020H09 gene |
Gene Aliases: |
ENSMUSG00000075426 |
Location: |
Chr.15:98567324-98570864 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9330020H09Rik | NR_028442.1 | |||
| AK140384.1 | BAE24365.1 |
Target Gene Details
Entrez Gene ID: | 12455 |
Gene Name: | cyclin T1 |
Gene Aliases: |
2810478G24Rik, AI115585, CycT1 |
Location: |
Chr.15:98543211-98568497 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnt1 | NM_009833.1 | NP_033963.1 | ||
| AF087662.1 | AAD17798.1 | |||
| AF095640.1 | AAD13656.1 | |||
| AF109179.1 | AAD19654.1 | |||
| AF113951.1 | AAD17205.1 | |||
| AK083037.1 | 1 | 81 | ||
| BC131685.1 |
More Information
Set Membership: |
|

Genomic Map