Assay Details
Target Gene Details
Entrez Gene ID: | 12455 |
Gene Name: | cyclin T1 |
Gene Aliases: |
2810478G24Rik, AI115585, CycT1 |
Location: |
Chr.15:98543211-98568497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnt1 | NM_009833.1 | NP_033963.1 | ||
| AF087662.1 | AAD17798.1 | |||
| AF095640.1 | AAD13656.1 | |||
| AF109179.1 | AAD19654.1 | |||
| AF113951.1 | AAD17205.1 | |||
| BC131685.1 |
More Information
Set Membership: |
|

Genomic Map