Assay Details
Target Gene Details
Entrez Gene ID: | 109754 |
Gene Name: | cytochrome b5 reductase 3 |
Gene Aliases: |
0610016L08Rik, 2500002N19Rik, C85115, Dia-1, Dia1, WU:AL591952.1-001, WU:AL591952.1-002, WU:AL591952.1-003, WU:Cyb5r3 |
Location: |
Chr.15:83153495-83172208 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyb5r3 | NM_029787.2 | NP_084063.1 | ||
| AF332059.1 | AAK56088.1 | |||
| AF332060.1 | AAK56089.1 | |||
| AK002640.1 | BAB22252.1 | |||
| AK010858.1 | BAB27227.1 | |||
| AK128917.1 | ||||
| AK177067.1 | ||||
| AK202011.1 | ||||
| AK208840.1 | ||||
| BC004760.1 | AAH04760.1 | |||
| BC032013.1 | AAH32013.1 | |||
| BC043074.1 | AAH43074.1 |
More Information
Set Membership: |
|

Genomic Map