Assay Details
Target Gene Details
Entrez Gene ID: | 215654 |
Gene Name: | cadherin 12 |
Gene Aliases: |
Cdhb |
Location: |
Chr.15:20448879-21597005 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh12 | NM_001008420.2 | NP_001008420.1 | ||
| BC086655.1 | AAH86655.1 |
More Information
Set Membership: |
|

Genomic Map