Assay Details
Target Gene Details
Entrez Gene ID: | 14904 |
Gene Name: | GTP binding protein 1 |
Gene Aliases: |
AL022987, AW045683, C85212, GP-1, Gtpbp |
Location: |
Chr.15:79690896-79721479 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp1 | NM_013818.2 | NP_038846.2 | ||
| AK004612.1 | BAB23409.1 | |||
| AK147717.1 | BAE28091.1 | |||
| AK150068.1 | BAE29280.1 | |||
| AK150185.1 | BAE29365.1 | |||
| AK158342.1 | ||||
| AK170091.1 | BAE41557.1 | |||
| BC046228.1 | AAH46228.1 | |||
| U87965.1 | AAB51274.1 |
More Information
Set Membership: |
|

Genomic Map