Assay Details
Target Gene Details
Entrez Gene ID: | 239554 |
Gene Name: | FAD-dependent oxidoreductase domain containing 2 |
Gene Aliases: |
A430097D04Rik, D15Bwg0759e, Gm304 |
Location: |
Chr.15:77940522-77956771 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxred2 | NM_001017983.2 | NP_001017983.2 | ||
| NM_001168260.1 | NP_001161732.1 | |||
| AK089177.1 | 1 | 148 | ||
| AK140027.1 | BAE24215.1 | |||
| AK170795.1 | BAE42033.1 | |||
| AK171308.1 | 1 | 148 | BAE42384.1 | |
| BC086662.1 | AAH86662.1 |
More Information
Set Membership: |
|

Genomic Map