Assay Details
Target Gene Details
Entrez Gene ID: | 74174 |
Gene Name: | gametocyte specific factor 1 |
Gene Aliases: |
1700006H03Rik, Cue110 |
Location: |
Chr.15:103402458-103430448 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtsf1 | NM_028797.1 | 4 | 287 | NP_083073.1 |
| AK005675.1 | 4 | 265 | BAB24181.1 | |
| BU945975.1 | 4 | 289 | ||
| CO812705.1 | 4 | 311 |

Genomic Map