Assay Details
Target Gene Details
Entrez Gene ID: | 12565 |
Gene Name: | cadherin 9 |
Gene Aliases: |
- |
Location: |
Chr.15:16728549-16857097 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh9 | NM_009869.1 | 5 | 868 | NP_033999.1 |
More Information
Set Membership: |
|

Genomic Map