Assay Details
Target Gene Details
Entrez Gene ID: | 109901 |
Gene Name: | chymotrypsin-like elastase family, member 1 |
Gene Aliases: |
1810009A17Rik, 1810062B19Rik, Ela-1, Ela1, PC-TsF |
Location: |
Chr.15:100674422-100687920 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cela1 | NM_033612.2 | NP_291090.2 | ||
| AK007392.1 | BAB25008.1 | |||
| BC011218.1 | AAH11218.1 | |||
| CB951068.1 | ||||
| M27347.1 | AAA39901.1 |
More Information
Set Membership: |
|

Genomic Map