Assay Details
Target Gene Details
Entrez Gene ID: | 76737 |
Gene Name: | cysteine-rich with EGF-like domains 2 |
Gene Aliases: |
5730592L21Rik, C85758 |
Location: |
Chr.15:88819646-88826681 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creld2 | NM_029720.2 | 1 | 50 | NP_083996.1 |
| AK017880.1 | 1 | 51 | BAB30986.1 | |
| AK165884.1 | 1 | 49 | BAE38436.1 | |
| BC047370.1 | AAH47370.1 |
More Information
Set Membership: |
|

Genomic Map