Assay Details
Target Gene Details
Entrez Gene ID: | 75705 |
Gene Name: | eukaryotic translation initiation factor 4B |
Gene Aliases: |
2310046H11Rik, AL024095, C85189, Eif4a2 |
Location: |
Chr.15:102073773-102097173 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4b | NM_145625.3 | NP_663600.2 | ||
| AK045250.1 | BAC32280.1 | |||
| AK077423.1 | BAC36793.1 | |||
| AK148012.1 | BAE28289.1 | |||
| AK161693.1 | BAE36536.1 | |||
| AK163266.1 | ||||
| AK166363.1 | BAE38733.1 | |||
| AK168350.1 | BAE40287.1 | |||
| AK168425.1 | ||||
| AK170256.1 | BAE41664.1 | |||
| AK170354.1 | BAE41740.1 | |||
| AK178737.1 | ||||
| AK179593.1 | ||||
| AK180578.1 | ||||
| AK186828.1 | ||||
| AK196954.1 | ||||
| AK206900.1 | ||||
| AK210063.1 | ||||
| AK211186.1 | ||||
| AK219884.1 | ||||
| BC007171.1 | AAH07171.1 |
More Information
Set Membership: |
|

Genomic Map