Assay Details
Target Gene Details
Entrez Gene ID: | 625599 |
Gene Name: | GPI anchored molecule like protein |
Gene Aliases: |
EG625599, HemT |
Location: |
Chr.15:74813435-74818815 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gml | NM_001177524.1 | 3 | 397 | NP_001170995.1 |
| AJ242831.1 | 2 | 456 | CAB57316.1 | |
| CJ042352.1 | 3 | 310 |

Genomic Map