Assay Details
Target Gene Details
Entrez Gene ID: | 70363 |
Gene Name: | family with sequence similarity 135, member B |
Gene Aliases: |
1700010C24Rik, A830008O07Rik |
Location: |
Chr.15:71438951-71728436 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam135b | NM_177819.3 | 6 | 604 | NP_808487.2 |
| AK043571.1 | 6 | 605 | BAC31584.1 | |
| BC125645.1 | 6 | 479 | ||
| BC125647.1 | 6 | 479 |

Genomic Map