Assay Details
Target Gene Details
Entrez Gene ID: | 105853 |
Gene Name: | mal, T cell differentiation protein 2 |
Gene Aliases: |
AI461653 |
Location: |
Chr.15:54571366-54602846 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mal2 | NM_178920.4 | NP_849251.1 | ||
| AK035986.1 | BAC29268.1 | |||
| BC113154.1 | AAI13155.1 |
More Information
Set Membership: |
|

Genomic Map