Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g6 | NM_001199023.1 | 1 | 160 | NP_001185952.1 |
| NM_001199024.1 | 1 | 160 | NP_001185953.1 | |
| NM_001199025.1 | NP_001185954.1 | |||
| NM_016915.4 | 1 | 160 | NP_058611.1 | |
| AK144495.1 | BAE25917.1 | |||
| AK153333.1 | BAE31912.1 | |||
| BC052845.1 | AAH52845.1 | |||
| BY099728.1 | ||||
| CF531790.1 |
More Information
Set Membership: |
|

Genomic Map