Assay Details
Target Gene Details
Entrez Gene ID: | 53357 |
Gene Name: | phospholipase A2, group VI |
Gene Aliases: |
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta |
Location: |
Chr.15:79286228-79328530 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g6 | NM_001199023.1 | 13 | 2557 | NP_001185952.1 |
| NM_001199024.1 | 12 | 2392 | NP_001185953.1 | |
| NM_001199025.1 | 13 | 1897 | NP_001185954.1 | |
| NM_016915.4 | 12 | 1938 | NP_058611.1 | |
| AF259401.1 | 12 | 1837 | AAF72651.1 | |
| AK144495.1 | 13 | 1771 | BAE25917.1 | |
| AK153333.1 | 12 | 2198 | BAE31912.1 | |
| BC003487.1 | 12 | 1731 | AAH03487.1 | |
| BC052845.1 | 12 | 1763 | AAH52845.1 | |
| BC057209.1 | 13 | 2150 | AAH57209.1 | |
| U88624.2 | 11 | 1614 | AAB48511.2 |
More Information
Set Membership: |
|

Genomic Map