Assay Details
Target Gene Details
Entrez Gene ID: | 109052 |
Gene Name: | keratin 75 |
Gene Aliases: |
4732468K03Rik, AA589387, K6hf, Krt2-6hf, Krtcap1 |
Location: |
Chr.15:101563343-101573904 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt75 | NM_133357.3 | NP_579935.1 | ||
| AF343088.1 | AAK17206.1 | |||
| AK028972.1 | BAC26221.1 | |||
| AK037233.1 | BAC29769.1 | |||
| AK076367.1 | BAC36315.1 | |||
| BC137935.1 | ||||
| BC137936.1 |

Genomic Map